Product Details

SNP ID
rs201976159
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:86428446 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGTCATAGTGACTGACTGGTATG[A/G]GGCACATGGAGATGATCCATACACC
Phenotype
MIM: 604003
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLCA2 PubMed Links

Gene Details

Gene
CLCA2
Gene Name
chloride channel accessory 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006536.5 515 Missense Mutation GAG,GGG E118G NP_006527.1
XM_011542448.2 515 Missense Mutation GAG,GGG E118G XP_011540750.1

View Full Product Details