Product Details

SNP ID
rs202041458
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:147907977 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAATGGGCGACTGGAGTTTCCTG[A/G]GGAACATCTTGGAGGAGGTGAATGA
Phenotype
MIM: 600897
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GJA8 PubMed Links

Gene Details

Gene
GJA8
Gene Name
gap junction protein alpha 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005267.4 822 Missense Mutation AGG,GGG R8G NP_005258.2
XM_011509416.1 822 Missense Mutation AGG,GGG R8G XP_011507718.1
XM_011509417.1 822 Missense Mutation AGG,GGG R8G XP_011507719.1

View Full Product Details