Product Details

SNP ID
rs202214253
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:46936744 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTGGAAGGCTGGGGTCAGCATCC[G/T]TCGATGCTGGAACCATGTCTGCCCA
Phenotype
MIM: 601310
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CYP4A11 PubMed Links

Gene Details

Gene
CYP4A11
Gene Name
cytochrome P450 family 4 subfamily A member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000778.3 Intron NP_000769.2
NM_001319155.1 Intron NP_001306084.1
XM_005270539.1 Intron XP_005270596.1
XM_017000464.1 Intron XP_016855953.1
XM_017000465.1 Intron XP_016855954.1

View Full Product Details