Product Details

SNP ID
rs202228024
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:155321857 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCCTGGGCCTGCACTTGTCCCG[C/T]CGTCCTGAGCTACAGGAGGGGCCTT
Phenotype
MIM: 134629
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FDPS PubMed Links

Gene Details

Gene
FDPS
Gene Name
farnesyl diphosphate synthase
There are no transcripts associated with this gene.

Gene
LOC105371451
Gene Name
uncharacterized LOC105371451
There are no transcripts associated with this gene.

Gene
RUSC1
Gene Name
RUN and SH3 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105203.1 313 Silent Mutation CGC,CGT R28R NP_001098673.1
NM_001105204.1 313 Silent Mutation CGC,CGT R28R NP_001098674.1
NM_001105205.1 313 Intron NP_001098675.1
NM_001278227.1 313 Intron NP_001265156.1
NM_001278228.1 313 Intron NP_001265157.1
NM_001278229.1 313 Intron NP_001265158.1
NM_001278230.1 313 Intron NP_001265159.1
NM_014328.4 313 Intron NP_055143.2
XM_006711254.1 313 Silent Mutation CGC,CGT R28R XP_006711317.1
XM_006711256.1 313 Silent Mutation CGC,CGT R28R XP_006711319.1
XM_006711257.1 313 Intron XP_006711320.1
XM_017000891.1 313 Silent Mutation CGC,CGT R28R XP_016856380.1
XM_017000892.1 313 Intron XP_016856381.1
Gene
RUSC1-AS1
Gene Name
RUSC1 antisense RNA 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039517.1 313 Intron NP_001034606.1

View Full Product Details