Product Details

SNP ID
rs201937405
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:167431685 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGTGAAATCCCCTGGCTGTTAG[C/T]GAGGGGGCAGGGCCTGCATGTGAAG
Phenotype
MIM: 186780 MIM: 164175
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD247 PubMed Links

Gene Details

Gene
CD247
Gene Name
CD247 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000734.3 710 Missense Mutation CAC,CGC H163R NP_000725.1
NM_198053.2 710 Missense Mutation CAC,CGC H164R NP_932170.1
XM_011510144.2 710 Missense Mutation CAC,CGC H172R XP_011508446.1
XM_011510145.1 710 Missense Mutation CAC,CGC H171R XP_011508447.1
XM_017002800.1 710 Missense Mutation CAC,CGC H195R XP_016858289.1
XM_017002801.1 710 Missense Mutation CAC,CGC H194R XP_016858290.1
Gene
POU2F1
Gene Name
POU class 2 homeobox 1
There are no transcripts associated with this gene.

View Full Product Details