Product Details
- SNP ID
-
rs202045082
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.1:52899184 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAAAACCCTCTCACCTCCGTTCCT[C/T]GGTCAATGGCTACTTTGCCCAGCCG
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ECHDC2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3216045] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ECHDC2
- Gene Name
- enoyl-CoA hydratase domain containing 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001198961.1 |
4739 |
Missense Mutation |
CAA,CGA |
Q248R |
NP_001185890.1 |
NM_001198962.1 |
4739 |
Intron |
|
|
NP_001185891.1 |
NM_001319958.1 |
4739 |
Missense Mutation |
CAA,CGA |
Q171R |
NP_001306887.1 |
NM_018281.3 |
4739 |
Missense Mutation |
CAA,CGA |
Q217R |
NP_060751.2 |
XM_011541709.2 |
4739 |
Missense Mutation |
CAA,CGA |
Q248R |
XP_011540011.1 |
XM_011541710.2 |
4739 |
Missense Mutation |
CAA,CGA |
Q248R |
XP_011540012.1 |
XM_011541711.2 |
4739 |
Missense Mutation |
CAA,CGA |
Q217R |
XP_011540013.1 |
XM_011541712.2 |
4739 |
Missense Mutation |
CAA,CGA |
Q248R |
XP_011540014.1 |
XM_011541713.2 |
4739 |
Missense Mutation |
CAA,CGA |
Q202R |
XP_011540015.1 |
XM_011541715.2 |
4739 |
Missense Mutation |
CAA,CGA |
Q202R |
XP_011540017.1 |
XM_011541719.2 |
4739 |
Missense Mutation |
CAA,CGA |
Q202R |
XP_011540021.1 |
XM_011541720.2 |
4739 |
Missense Mutation |
CAA,CGA |
Q202R |
XP_011540022.1 |
XM_011541721.2 |
4739 |
Missense Mutation |
CAA,CGA |
Q202R |
XP_011540023.1 |
XM_011541722.2 |
4739 |
Missense Mutation |
CAA,CGA |
Q202R |
XP_011540024.1 |
XM_011541723.2 |
4739 |
Missense Mutation |
CAA,CGA |
Q202R |
XP_011540025.1 |
XM_011541724.2 |
4739 |
Missense Mutation |
CAA,CGA |
Q198R |
XP_011540026.1 |
XM_011541725.2 |
4739 |
Missense Mutation |
CAA,CGA |
Q217R |
XP_011540027.1 |
XM_011541726.2 |
4739 |
Missense Mutation |
CAA,CGA |
Q171R |
XP_011540028.1 |
XM_011541727.2 |
4739 |
Missense Mutation |
CAA,CGA |
Q152R |
XP_011540029.1 |
XM_017001638.1 |
4739 |
Missense Mutation |
CAA,CGA |
Q202R |
XP_016857127.1 |
XM_017001639.1 |
4739 |
Missense Mutation |
CAA,CGA |
Q202R |
XP_016857128.1 |
XM_017001640.1 |
4739 |
Missense Mutation |
CAA,CGA |
Q202R |
XP_016857129.1 |
XM_017001641.1 |
4739 |
Missense Mutation |
CAA,CGA |
Q202R |
XP_016857130.1 |
XM_017001642.1 |
4739 |
Missense Mutation |
CAA,CGA |
Q171R |
XP_016857131.1 |
- Gene
- ZYG11A
- Gene Name
- zyg-11 family member A, cell cycle regulator
There are no transcripts associated with this gene.
View Full Product Details