Product Details
- SNP ID
-
rs201575332
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:96049 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGTTGGAAGAATAATGTAGGACAT[C/T]GCAGAAGACGATGTTTAGATACTGA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DEFB125
PubMed Links
Gene Details
- Gene
- DEFB125
- Gene Name
- defensin beta 125
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_153325.3 |
141 |
Missense Mutation |
CGC,TGC |
R35C |
NP_697020.2 |
View Full Product Details