Product Details

SNP ID
rs201814399
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:35547434 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAGCTTGGGAAAGTCGAGGTGAC[A/G]GTGTTTGACCCTGACTCCCTGGACC
Phenotype
MIM: 616971
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ERGIC3 PubMed Links

Gene Details

Gene
ERGIC3
Gene Name
ERGIC and golgi 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015966.2 459 Silent Mutation ACA,ACG T130T NP_057050.1
NM_198398.1 459 Silent Mutation ACA,ACG T130T NP_938408.1

View Full Product Details