Product Details

SNP ID
rs201864662
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:5547686 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAAGCATTCTCCACGTTATCAATG[C/T]CGTTGGCATCCACATGGATATCAGA
Phenotype
MIM: 614124
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
GPCPD1 PubMed Links

Gene Details

Gene
GPCPD1
Gene Name
glycerophosphocholine phosphodiesterase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019593.3 2358 Missense Mutation GAC,GGC D665G NP_062539.1
XM_005260758.4 2358 Missense Mutation GAC,GGC D665G XP_005260815.1
XM_005260761.1 2358 Missense Mutation GAC,GGC D484G XP_005260818.1
XM_006723596.3 2358 Missense Mutation GAC,GGC D665G XP_006723659.1

View Full Product Details