Product Details

SNP ID
rs199658514
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:34929527 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTCCATGAGGATGTAGGAGGCC[A/C]TCTCCTCACTGTCCTTCAGCTGTTT
Phenotype
MIM: 605832 MIM: 601002
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ACSS2 PubMed Links

Gene Details

Gene
ACSS2
Gene Name
acyl-CoA synthetase short-chain family member 2
There are no transcripts associated with this gene.

Gene
GSS
Gene Name
glutathione synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000178.3 1274 Missense Mutation AGG,ATG R392M NP_000169.1
NM_001322494.1 1274 Missense Mutation AGG,ATG R392M NP_001309423.1
NM_001322495.1 1274 Missense Mutation AGG,ATG R392M NP_001309424.1

View Full Product Details