Product Details
- SNP ID
-
rs200972775
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:31968408 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GACGGTGCCACGGACCTGTGGCTGG[C/G]GGCCTCCTACTACCTGCAGAATCAA
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
XKR7
PubMed Links
Gene Details
- Gene
- XKR7
- Gene Name
- XK related 7
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001011718.1 |
407 |
Missense Mutation |
GCG,GGG |
A78G |
NP_001011718.1 |
View Full Product Details