Product Details

SNP ID
rs201380485
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:35279739 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATCACCTCACTGCCTCGGTTCAC[A/G]GTCCCCGCCTGCCAAGGGATGGGCT
Phenotype
MIM: 602912 MIM: 604871
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EIF6 PubMed Links

Gene Details

Gene
EIF6
Gene Name
eukaryotic translation initiation factor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267810.1 785 Silent Mutation ACC,ACT T185T NP_001254739.1
NM_002212.3 785 Silent Mutation ACC,ACT T185T NP_002203.1
NM_181466.2 785 Silent Mutation ACC,ACT T166T NP_852131.1
NM_181468.2 785 Silent Mutation ACC,ACT T185T NP_852133.1
Gene
FAM83C
Gene Name
family with sequence similarity 83 member C
There are no transcripts associated with this gene.

Gene
FAM83C-AS1
Gene Name
FAM83C antisense RNA 1
There are no transcripts associated with this gene.

Gene
MMP24
Gene Name
matrix metallopeptidase 24
There are no transcripts associated with this gene.

Gene
MMP24-AS1
Gene Name
MMP24 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details