Product Details

SNP ID
rs200823009
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:3754400 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAGGAGCCTCACGCTCTCACCCA[C/T]GACGCGGGCCTGCACCGTCACGCGC
Phenotype
MIM: 610702
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C20orf27 PubMed Links

Gene Details

Gene
C20orf27
Gene Name
chromosome 20 open reading frame 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039140.2 1104 Missense Mutation NP_001034229.1
NM_001258429.1 1104 Missense Mutation NP_001245358.1
NM_001258430.1 1104 Missense Mutation NP_001245359.1
XM_011529266.2 1104 Intron XP_011527568.1
Gene
HSPA12B
Gene Name
heat shock protein family A (Hsp70) member 12B
There are no transcripts associated with this gene.

View Full Product Details