Product Details

SNP ID
rs200471798
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:58460858 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGGAAGGGGGCTATGTGAAGAC[A/C]CCCTGTGCTGGGGTGCTTCTGCAGC
Phenotype
MIM: 605704
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
APCDD1L PubMed Links

Gene Details

Gene
APCDD1L
Gene Name
APC down-regulated 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304787.1 1669 Missense Mutation GGT,TGT G491C NP_001291716.1
NM_153360.2 1669 Missense Mutation GGT,TGT G480C NP_699191.1
Gene
VAPB
Gene Name
VAMP associated protein B and C
There are no transcripts associated with this gene.

View Full Product Details