Product Details

SNP ID
rs199718150
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45630963 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGAATCTCACCGAGATTGTCAAG[A/C]AAATAACATATGCTGTTCTACCTAC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
WFDC10A PubMed Links

Gene Details

Gene
WFDC10A
Gene Name
WAP four-disulfide core domain 10A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080753.2 167 Intron NP_542791.1
XM_017027679.1 167 Missense Mutation GAA,GCA E56A XP_016883168.1
Gene
WFDC9
Gene Name
WAP four-disulfide core domain 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_147198.3 167 Intron NP_671731.1

View Full Product Details