Product Details

SNP ID
rs200377875
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:35174924 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTCCAGTTCCACGGCCTCGTGC[G/T]CCTGGTGCACCAGGAGCGGACCTTG
Phenotype
MIM: 600646
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PROCR PubMed Links

Gene Details

Gene
PROCR
Gene Name
protein C receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006404.4 511 Missense Mutation CGC,CTC R98L NP_006395.2
XM_011528496.1 511 Missense Mutation CGC,CTC R98L XP_011526798.1

View Full Product Details