Product Details

SNP ID
rs200828167
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:31844761 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGAGGTGCTCAAGGCCTGGGTCA[A/G]CCCCCATGCAAAAATTCAGTGCCTG
Phenotype
MIM: 616776 MIM: 602939
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DUSP15 PubMed Links

Gene Details

Gene
DUSP15
Gene Name
dual specificity phosphatase 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012644.2 857 Intron NP_001012662.1
NM_001320478.1 857 Intron NP_001307407.1
NM_001320479.1 857 Intron NP_001307408.1
NM_080611.4 857 Intron NP_542178.2
NM_177991.2 857 Intron NP_817130.1
XM_017027654.1 857 Intron XP_016883143.1
XM_017027655.1 857 Intron XP_016883144.1
XM_017027656.1 857 Intron XP_016883145.1
XM_017027657.1 857 Intron XP_016883146.1
XM_017027658.1 857 Intron XP_016883147.1
XM_017027659.1 857 Intron XP_016883148.1
XM_017027660.1 857 Intron XP_016883149.1
XM_017027661.1 857 Intron XP_016883150.1
XM_017027662.1 857 Intron XP_016883151.1
Gene
FOXS1
Gene Name
forkhead box S1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004118.3 857 Missense Mutation GCT,GTT A261V NP_004109.1

View Full Product Details