Product Details

SNP ID
rs200830479
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:34845344 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGCTGCTCTACAGGATGGTGGCT[C/G]CAGCTGCATCTGGGCTCCGAGGGTC
Phenotype
MIM: 612342
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GGT7 PubMed Links

Gene Details

Gene
GGT7
Gene Name
gamma-glutamyltransferase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178026.2 1495 Missense Mutation GCA,GGA A658G NP_821158.2
XM_011528782.2 1495 Missense Mutation GCA,GGA A424G XP_011527084.1
XM_011528783.2 1495 Missense Mutation GCA,GGA A375G XP_011527085.1
XM_017027809.1 1495 UTR 3 XP_016883298.1
XM_017027810.1 1495 Intron XP_016883299.1
XM_017027811.1 1495 Intron XP_016883300.1

View Full Product Details