Product Details

SNP ID
rs200178576
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:62216180 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTTTCGTACCAGTAGTCAGGGAC[G/T]CAGTGGCCATGGCAGGCGGCCCGGA
Phenotype
MIM: 604525
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HRH3 PubMed Links

Gene Details

Gene
HRH3
Gene Name
histamine receptor H3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007232.2 1164 Silent Mutation NP_009163.2
XM_005260266.3 1164 Silent Mutation XP_005260323.1
XM_017027623.1 1164 Silent Mutation XP_016883112.1

View Full Product Details