Product Details

SNP ID
rs200630434
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63862380 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGACGACCTTGTGCCGCTGGCGCT[G/T]AAGGTCATGCCCCACAGTTGGAAGG
Phenotype
MIM: 603747
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
ABHD16B PubMed Links

Gene Details

Gene
ABHD16B
Gene Name
abhydrolase domain containing 16B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080622.3 1168 Silent Mutation CTG,CTT L280L NP_542189.1
Gene
TPD52L2
Gene Name
tumor protein D52 like 2
There are no transcripts associated with this gene.

View Full Product Details