Product Details

SNP ID
rs200883545
Assay Type
Functionally Tested
NCBI dbSNP Submissions
4
Location
Chr.20:63540929 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCATGAGCACGTAGACCTCCGC[C/T]GGGCAGGCAGCCGGGCGCGGCAGCC
Phenotype
MIM: 602004
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
PTK6 PubMed Links

Gene Details

Gene
PTK6
Gene Name
protein tyrosine kinase 6
There are no transcripts associated with this gene.

Gene
SRMS
Gene Name
src-related kinase lacking C-terminal regulatory tyrosine and N-terminal myristylation sites
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080823.3 1397 Silent Mutation CCA,CCG P452P NP_543013.1

View Full Product Details