Product Details

SNP ID
rs201832808
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63939996 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGCTAACCCGTGTAGGCCACTT[C/T]CTCCTCGTCACTGCCATCGGGGACC
Phenotype
MIM: 611203 MIM: 610866
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJC5 PubMed Links

Gene Details

Gene
DNAJC5
Gene Name
DnaJ heat shock protein family (Hsp40) member C5
There are no transcripts associated with this gene.

Gene
MIR1914
Gene Name
microRNA 1914
There are no transcripts associated with this gene.

Gene
MIR647
Gene Name
microRNA 647
There are no transcripts associated with this gene.

Gene
UCKL1
Gene Name
uridine-cytidine kinase 1 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193379.1 1908 Missense Mutation AAA,GAA K528E NP_001180308.1
NM_017859.3 1908 Missense Mutation AAA,GAA K543E NP_060329.2
XM_005260216.2 1908 Missense Mutation AAA,GAA K551E XP_005260273.1
XM_006723806.2 1908 Missense Mutation AAA,GAA K553E XP_006723869.1
XM_006723807.2 1908 Missense Mutation AAA,GAA K552E XP_006723870.1
XM_006723808.1 1908 Missense Mutation AAA,GAA K552E XP_006723871.1
XM_006723809.1 1908 Missense Mutation AAA,GAA K545E XP_006723872.1
XM_006723810.1 1908 Missense Mutation AAA,GAA K544E XP_006723873.1
XM_006723811.3 1908 Missense Mutation AAA,GAA K537E XP_006723874.1
XM_011528868.2 1908 Missense Mutation AAA,GAA K538E XP_011527170.1
XM_011528869.1 1908 Missense Mutation AAA,GAA K437E XP_011527171.1
XM_011528870.1 1908 Missense Mutation AAA,GAA K427E XP_011527172.1
XM_011528871.1 1908 Missense Mutation AAA,GAA K427E XP_011527173.1
XM_011528872.1 1908 Missense Mutation AAA,GAA K427E XP_011527174.1
XM_011528873.2 1908 Intron XP_011527175.1
XM_017027894.1 1908 Intron XP_016883383.1
XM_017027895.1 1908 Intron XP_016883384.1
XM_017027896.1 1908 Missense Mutation AAA,GAA K544E XP_016883385.1
XM_017027897.1 1908 Intron XP_016883386.1
XM_017027898.1 1908 Missense Mutation AAA,GAA K543E XP_016883387.1
XM_017027899.1 1908 Missense Mutation AAA,GAA K542E XP_016883388.1
XM_017027900.1 1908 Missense Mutation AAA,GAA K528E XP_016883389.1
XM_017027901.1 1908 Missense Mutation AAA,GAA K527E XP_016883390.1
XM_017027902.1 1908 Missense Mutation AAA,GAA K429E XP_016883391.1
XM_017027903.1 1908 Missense Mutation AAA,GAA K425E XP_016883392.1
XM_017027904.1 1908 Missense Mutation AAA,GAA K337E XP_016883393.1
XM_017027905.1 1908 Missense Mutation AAA,GAA K335E XP_016883394.1
XM_017027906.1 1908 Missense Mutation AAA,GAA K335E XP_016883395.1
XM_017027907.1 1908 Missense Mutation AAA,GAA K324E XP_016883396.1
XM_017027908.1 1908 Missense Mutation AAA,GAA K324E XP_016883397.1

View Full Product Details