Product Details

SNP ID
rs201983158
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:31819586 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCACACGCCTCCCTACCTCATGGC[A/G]ACAGAAAATGGAGCAGTTGAGCTGG
Phenotype
MIM: 606566
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MYLK2 PubMed Links

Gene Details

Gene
MYLK2
Gene Name
myosin light chain kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033118.3 108 Silent Mutation GCA,GCG A2A NP_149109.1

View Full Product Details