Product Details
- SNP ID
-
rs202092326
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:62306211 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCTCTTGCAGGAACCCAAGACAGA[C/G]CAGGATGAGGAGCATTGCCGGAAAG
- Phenotype
-
MIM: 610650
MIM: 601033
MIM: 606731
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ADRM1
PubMed Links
Gene Details
- Gene
- ADRM1
- Gene Name
- adhesion regulating molecule 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001281437.1 |
1335 |
Missense Mutation |
GAC,GAG |
D76E |
NP_001268366.1 |
NM_001281438.1 |
1335 |
Missense Mutation |
GAC,GAG |
D76E |
NP_001268367.1 |
NM_007002.3 |
1335 |
Missense Mutation |
GAC,GAG |
D115E |
NP_008933.2 |
NM_175573.2 |
1335 |
Missense Mutation |
GAC,GAG |
D115E |
NP_783163.1 |
XM_005260257.1 |
1335 |
Missense Mutation |
GAC,GAG |
D115E |
XP_005260314.1 |
XM_011528503.1 |
1335 |
Missense Mutation |
GAC,GAG |
D115E |
XP_011526805.1 |
XM_017027602.1 |
1335 |
Missense Mutation |
GAC,GAG |
D115E |
XP_016883091.1 |
XM_017027603.1 |
1335 |
Missense Mutation |
GAC,GAG |
D115E |
XP_016883092.1 |
XM_017027604.1 |
1335 |
Missense Mutation |
GAC,GAG |
D115E |
XP_016883093.1 |
XM_017027605.1 |
1335 |
Missense Mutation |
GAC,GAG |
D115E |
XP_016883094.1 |
XM_017027606.1 |
1335 |
Missense Mutation |
GAC,GAG |
D76E |
XP_016883095.1 |
XM_017027607.1 |
1335 |
Missense Mutation |
GAC,GAG |
D76E |
XP_016883096.1 |
XM_017027608.1 |
1335 |
UTR 5 |
|
|
XP_016883097.1 |
XM_017027609.1 |
1335 |
UTR 5 |
|
|
XP_016883098.1 |
XM_017027610.1 |
1335 |
UTR 5 |
|
|
XP_016883099.1 |
- Gene
- LAMA5
- Gene Name
- laminin subunit alpha 5
There are no transcripts associated with this gene.
- Gene
- OSBPL2
- Gene Name
- oxysterol binding protein like 2
There are no transcripts associated with this gene.
View Full Product Details