Product Details

SNP ID
rs199984812
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:31844796 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAATTCAGTGCCTGCAGCCGACA[C/T]TGGTAGCTGCTCCCGATGCCTGATT
Phenotype
MIM: 616776 MIM: 602939
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DUSP15 PubMed Links

Gene Details

Gene
DUSP15
Gene Name
dual specificity phosphatase 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012644.2 822 Intron NP_001012662.1
NM_001320478.1 822 Intron NP_001307407.1
NM_001320479.1 822 Intron NP_001307408.1
NM_080611.4 822 Intron NP_542178.2
NM_177991.2 822 Intron NP_817130.1
XM_017027654.1 822 Intron XP_016883143.1
XM_017027655.1 822 Intron XP_016883144.1
XM_017027656.1 822 Intron XP_016883145.1
XM_017027657.1 822 Intron XP_016883146.1
XM_017027658.1 822 Intron XP_016883147.1
XM_017027659.1 822 Intron XP_016883148.1
XM_017027660.1 822 Intron XP_016883149.1
XM_017027661.1 822 Intron XP_016883150.1
XM_017027662.1 822 Intron XP_016883151.1
Gene
FOXS1
Gene Name
forkhead box S1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004118.3 822 Silent Mutation CAA,CAG Q249Q NP_004109.1

View Full Product Details