Product Details

SNP ID
rs201204270
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:31665985 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGAGTGAGCCCAGCAGAACCAC[A/G]CCGGCCACAGTCATGCCCGTCAGGA
Phenotype
MIM: 600039
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BCL2L1 PubMed Links

Gene Details

Gene
BCL2L1
Gene Name
BCL2 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001191.3 1043 Silent Mutation GGC,GGT G159G NP_001182.1
NM_001317919.1 1043 Silent Mutation GGC,GGT G222G NP_001304848.1
NM_001317920.1 1043 Silent Mutation GGC,GGT G222G NP_001304849.1
NM_001317921.1 1043 Silent Mutation GGC,GGT G222G NP_001304850.1
NM_001322239.1 1043 Silent Mutation GGC,GGT G222G NP_001309168.1
NM_001322240.1 1043 Silent Mutation GGC,GGT G222G NP_001309169.1
NM_001322242.1 1043 Silent Mutation GGC,GGT G222G NP_001309171.1
NM_138578.2 1043 Silent Mutation GGC,GGT G222G NP_612815.1
XM_011528964.2 1043 Silent Mutation GGC,GGT G222G XP_011527266.1
XM_017027993.1 1043 Silent Mutation GGC,GGT G222G XP_016883482.1

View Full Product Details