Product Details
- SNP ID
-
rs201204862
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:1980397 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGAGTCACCACCTTGAACTGGCGCC[G/T]GAGAAAACCGCCATAGCGCTTCTGG
- Phenotype
-
MIM: 131340
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
LOC727993
PubMed Links
Gene Details
- Gene
- LOC727993
- Gene Name
- uncharacterized LOC727993
There are no transcripts associated with this gene.
- Gene
- PDYN
- Gene Name
- prodynorphin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001190892.1 |
966 |
Silent Mutation |
GGG,TGG |
G231W |
NP_001177821.1 |
NM_001190898.2 |
966 |
Silent Mutation |
GGG,TGG |
G231W |
NP_001177827.1 |
NM_001190899.2 |
966 |
Silent Mutation |
GGG,TGG |
G231W |
NP_001177828.1 |
NM_001190900.1 |
966 |
Silent Mutation |
GGG,TGG |
G231W |
NP_001177829.1 |
NM_024411.4 |
966 |
Silent Mutation |
GGG,TGG |
G231W |
NP_077722.1 |
XM_011529244.1 |
966 |
Missense Mutation |
GGG,TGG |
G231W |
XP_011527546.1 |
XM_011529245.1 |
966 |
Missense Mutation |
GGG,TGG |
G231W |
XP_011527547.1 |
XM_011529246.2 |
966 |
Missense Mutation |
GGG,TGG |
G231W |
XP_011527548.1 |
XM_011529247.1 |
966 |
Missense Mutation |
GGG,TGG |
G231W |
XP_011527549.1 |
XM_011529248.1 |
966 |
Missense Mutation |
GGG,TGG |
G231W |
XP_011527550.1 |
XM_011529249.2 |
966 |
Silent Mutation |
GGG,TGG |
G231W |
XP_011527551.1 |
XM_011529250.2 |
966 |
Missense Mutation |
GGG,TGG |
G231W |
XP_011527552.1 |
XM_017027878.1 |
966 |
Missense Mutation |
GGG,TGG |
G231W |
XP_016883367.1 |
View Full Product Details