Product Details

SNP ID
rs201304064
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63643865 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGCTCTCTGGGGACAGGTCAGAA[A/G]GGGACTTGAGGATGACCTCGAAGCT
Phenotype
MIM: 608362
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
STMN3 PubMed Links

Gene Details

Gene
STMN3
Gene Name
stathmin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276310.1 447 Missense Mutation CCT,CTT P50L NP_001263239.1
NM_015894.3 447 Missense Mutation CCT,CTT P61L NP_056978.2

View Full Product Details