Product Details

SNP ID
rs200038988
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:25248355 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTACTTCTTCGCGCTGGCGCACAC[G/T]GTGCGCGACCACCTCGTGGGCCGCT
Phenotype
MIM: 138550
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC105372580 PubMed Links

Gene Details

Gene
LOC105372580
Gene Name
uncharacterized LOC105372580
There are no transcripts associated with this gene.

Gene
PYGB
Gene Name
phosphorylase, glycogen; brain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002862.3 286 Silent Mutation ACG,ACT T59T NP_002853.2

View Full Product Details