Product Details

SNP ID
rs200696786
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45815393 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTAAAAAAACTTTTTAAAAAGGCA[C/G]TGGGGAGCATCAGAACCAGCTCAAC
Phenotype
MIM: 611388 MIM: 191039 MIM: 605574
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
DNTTIP1 PubMed Links

Gene Details

Gene
DNTTIP1
Gene Name
deoxynucleotidyltransferase, terminal, interacting protein 1
There are no transcripts associated with this gene.

Gene
TNNC2
Gene Name
troponin C2, fast skeletal type
There are no transcripts associated with this gene.

Gene
UBE2C
Gene Name
ubiquitin conjugating enzyme E2 C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281741.1 Intron NP_001268670.1
NM_001281742.1 Intron NP_001268671.1
NM_007019.3 Intron NP_008950.1
NM_181799.2 Intron NP_861515.1
NM_181800.2 Intron NP_861516.1
NM_181801.3 Intron NP_861517.1

View Full Product Details