Product Details

SNP ID
rs199790941
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:23036120 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGGCCTGGTCGGCAGTCTTCGTG[G/T]TCTACACTTTCCTGCTGGGCTTCCT
Phenotype
MIM: 182454 MIM: 188040
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC101929707 PubMed Links

Gene Details

Gene
LOC101929707
Gene Name
uncharacterized LOC101929707
There are no transcripts associated with this gene.

Gene
SSTR4
Gene Name
somatostatin receptor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001052.2 701 Missense Mutation GTC,TTC V213F NP_001043.2
Gene
THBD
Gene Name
thrombomodulin
There are no transcripts associated with this gene.

View Full Product Details