Product Details

SNP ID
rs199791180
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:31820299 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAACCCTCAACTAGCAGCCAAGGC[C/T]CCAAAGGAGAGGGTGACAGGGGCGG
Phenotype
MIM: 606566
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYLK2 PubMed Links

Gene Details

Gene
MYLK2
Gene Name
myosin light chain kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033118.3 328 Missense Mutation CCC,TCC P76S NP_149109.1

View Full Product Details