Product Details

SNP ID
rs202001008
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:25675385 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATTTCCCTTCCAGTTGAAGCCTC[A/G]CCCACACTCCTTGCATACAAAAGGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF337 PubMed Links

Gene Details

Gene
ZNF337
Gene Name
zinc finger protein 337
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290261.1 1774 Nonsense Mutation CGA,TGA R635* NP_001277190.1
NM_015655.3 1774 Nonsense Mutation CGA,TGA R635* NP_056470.1
XM_005260702.3 1774 Nonsense Mutation CGA,TGA R603* XP_005260759.1
XM_006723558.3 1774 Nonsense Mutation CGA,TGA R635* XP_006723621.1
XM_011529219.2 1774 Nonsense Mutation CGA,TGA R635* XP_011527521.1
XM_017027802.1 1774 Nonsense Mutation CGA,TGA R603* XP_016883291.1
XM_017027803.1 1774 Nonsense Mutation CGA,TGA R603* XP_016883292.1
XM_017027804.1 1774 Nonsense Mutation CGA,TGA R570* XP_016883293.1
Gene
ZNF337-AS1
Gene Name
ZNF337 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details