Product Details

SNP ID
rs200252509
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45939046 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCCCCGGGAGGAAGCGTCCCTGC[C/T]GAGTCACTCCCCAGGTACCTCCAAT
Phenotype
MIM: 610827
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC107985388 PubMed Links

Gene Details

Gene
LOC107985388
Gene Name
pre-mRNA-splicing factor cwc22-like
There are no transcripts associated with this gene.

Gene
PCIF1
Gene Name
PDX1 C-terminal inhibiting factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022104.3 325 Missense Mutation CCG,CTG P16L NP_071387.1
XM_011528980.2 325 Missense Mutation CCG,CTG P16L XP_011527282.1
XM_011528981.2 325 Missense Mutation CCG,CTG P16L XP_011527283.1
XM_017028013.1 325 Missense Mutation CCG,CTG P16L XP_016883502.1
XM_017028014.1 325 UTR 5 XP_016883503.1
Gene
ZNF335
Gene Name
zinc finger protein 335
There are no transcripts associated with this gene.

View Full Product Details