Product Details
- SNP ID
-
rs201516661
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:24949524 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCAGCCATGCGAGCGGCTGGAACT[C/T]TGCTGGCCTTCTGCTGCCTGGTCTT
- Phenotype
-
MIM: 603253
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CST7
PubMed Links
Gene Details
- Gene
- CST7
- Gene Name
- cystatin F
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003650.3 |
295 |
Silent Mutation |
CTG,TTG |
L7L |
NP_003641.3 |
View Full Product Details