Product Details

SNP ID
rs201517755
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:31844921 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCATGAGGAAGATGAGGTGGCCA[C/T]GGGGAGCTCCCCTGGGCATGCAGGC
Phenotype
MIM: 616776 MIM: 602939
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DUSP15 PubMed Links

Gene Details

Gene
DUSP15
Gene Name
dual specificity phosphatase 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012644.2 697 Intron NP_001012662.1
NM_001320478.1 697 Intron NP_001307407.1
NM_001320479.1 697 Intron NP_001307408.1
NM_080611.4 697 Intron NP_542178.2
NM_177991.2 697 Intron NP_817130.1
XM_017027654.1 697 Intron XP_016883143.1
XM_017027655.1 697 Intron XP_016883144.1
XM_017027656.1 697 Intron XP_016883145.1
XM_017027657.1 697 Intron XP_016883146.1
XM_017027658.1 697 Intron XP_016883147.1
XM_017027659.1 697 Intron XP_016883148.1
XM_017027660.1 697 Intron XP_016883149.1
XM_017027661.1 697 Intron XP_016883150.1
XM_017027662.1 697 Intron XP_016883151.1
Gene
FOXS1
Gene Name
forkhead box S1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004118.3 697 Missense Mutation NP_004109.1

View Full Product Details