Product Details

SNP ID
rs201016960
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:50959765 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCGGGGAACGGCCCACTGTGACTG[A/G]TCTGCTGGACTATGAAGCCTTCTGT
Phenotype
MIM: 603503 MIM: 609277
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DPM1 PubMed Links

Gene Details

Gene
DPM1
Gene Name
dolichyl-phosphate mannosyltransferase polypeptide 1, catalytic subunit
There are no transcripts associated with this gene.

Gene
MOCS3
Gene Name
molybdenum cofactor synthesis 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014484.4 952 Missense Mutation GAT,GGT D308G NP_055299.1

View Full Product Details