Product Details

SNP ID
rs199524771
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:38586129 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGGACACTCCACGGCCAAGAGAA[G/T]GAGAGGCCCTGCTGGTGAGCCTGCT
Phenotype
MIM: 611396
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ADIG PubMed Links

Gene Details

Gene
ADIG
Gene Name
adipogenin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018082.2 281 Missense Mutation AAG,AAT K75N NP_001018092.1
Gene
RALGAPB
Gene Name
Ral GTPase activating protein non-catalytic beta subunit
There are no transcripts associated with this gene.

View Full Product Details