Product Details

SNP ID
rs199529717
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:18024672 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCTCCTCACTCAGGCTGGTATT[C/T]TCCTGGTGTGCGGATGTCAGCTTGC
Phenotype
MIM: 616441
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OVOL2 PubMed Links

Gene Details

Gene
OVOL2
Gene Name
ovo like zinc finger 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303461.1 1040 Silent Mutation GAA,GAG E132E NP_001290390.1
NM_001303462.1 1040 Silent Mutation GAA,GAG E132E NP_001290391.1
NM_021220.3 1040 Silent Mutation GAA,GAG E264E NP_067043.2

View Full Product Details