Product Details

SNP ID
rs199532746
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:18142690 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGTAGCATCCACCTGAGTAGTCT[A/G]ATCAGTCGGCATGATGACGAAGCCA
Phenotype
MIM: 614771
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KAT14 PubMed Links

Gene Details

Gene
KAT14
Gene Name
lysine acetyltransferase 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020536.4 467 Silent Mutation CTA,CTG L10L NP_065397.3
Gene
PET117
Gene Name
PET117 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164811.1 467 UTR 3 NP_001158283.1

View Full Product Details