Product Details

SNP ID
rs199534645
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:58840470 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGACCGAGTCCGAAATCGAGTCC[A/G]AGACCGACTTCGAGACCGAGCCTGA
Phenotype
MIM: 139320 MIM: 610540
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GNAS PubMed Links

Gene Details

Gene
GNAS
Gene Name
GNAS complex locus
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000516.5 790 Intron NP_000507.1
NM_001077488.3 790 Intron NP_001070956.1
NM_001077489.3 790 Intron NP_001070957.1
NM_001077490.2 790 Intron NP_001070958.1
NM_001309840.1 790 Intron NP_001296769.1
NM_001309842.1 790 Intron NP_001296771.1
NM_001309861.1 790 Intron NP_001296790.1
NM_001309883.1 790 Intron NP_001296812.1
NM_016592.3 790 Missense Mutation AAG,GAG K122E NP_057676.1
NM_080425.3 790 Intron NP_536350.2
NM_080426.3 790 Intron NP_536351.1
XM_017027812.1 790 Intron XP_016883301.1
XM_017027813.1 790 Intron XP_016883302.1
XM_017027814.1 790 Intron XP_016883303.1
XM_017027815.1 790 Intron XP_016883304.1
XM_017027816.1 790 Intron XP_016883305.1
XM_017027817.1 790 Intron XP_016883306.1
XM_017027818.1 790 Intron XP_016883307.1
XM_017027819.1 790 Intron XP_016883308.1
XM_017027820.1 790 Intron XP_016883309.1
XM_017027821.1 790 Missense Mutation AAG,GAG K122E XP_016883310.1
XM_017027822.1 790 Missense Mutation AAG,GAG K122E XP_016883311.1
Gene
GNAS-AS1
Gene Name
GNAS antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details