Product Details

SNP ID
rs201921227
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:31038278 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTAGAAGGCAGAGAATCCATATC[A/C]TCCGTAGTATAATGGTCGGCAACAG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KRTAP19-8 PubMed Links

Gene Details

Gene
KRTAP19-8
Gene Name
keratin associated protein 19-8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099219.1 199 Nonsense Mutation GGA,TGA G56* NP_001092689.1

View Full Product Details