Product Details
- SNP ID
-
rs201880764
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:44012513 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACGCCCATGGACGCCTCTGAGGAGC[C/T]GCTGCCGCCGGTGATCTACACCATG
- Phenotype
-
MIM: 602103
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TRAPPC10
PubMed Links
Gene Details
- Gene
- TRAPPC10
- Gene Name
- trafficking protein particle complex 10
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003274.4 |
182 |
Missense Mutation |
CCG,CTG |
P7L |
NP_003265.3 |
XM_005261168.4 |
182 |
Missense Mutation |
CCG,CTG |
P7L |
XP_005261225.1 |
XM_011529714.2 |
182 |
Missense Mutation |
CCG,CTG |
P7L |
XP_011528016.1 |
XM_011529715.2 |
182 |
Missense Mutation |
CCG,CTG |
P7L |
XP_011528017.1 |
XM_011529716.2 |
182 |
Missense Mutation |
CCG,CTG |
P7L |
XP_011528018.1 |
XM_011529717.2 |
182 |
Missense Mutation |
CCG,CTG |
P7L |
XP_011528019.1 |
XM_011529718.2 |
182 |
Missense Mutation |
CCG,CTG |
P7L |
XP_011528020.1 |
XM_011529719.2 |
182 |
Missense Mutation |
CCG,CTG |
P7L |
XP_011528021.1 |
XM_011529720.2 |
182 |
Missense Mutation |
CCG,CTG |
P7L |
XP_011528022.1 |
XM_011529721.2 |
182 |
Intron |
|
|
XP_011528023.1 |
XM_011529722.2 |
182 |
Intron |
|
|
XP_011528024.1 |
XM_011529723.2 |
182 |
Intron |
|
|
XP_011528025.1 |
XM_011529725.2 |
182 |
Missense Mutation |
CCG,CTG |
P7L |
XP_011528027.1 |
XM_011529726.2 |
182 |
Missense Mutation |
CCG,CTG |
P7L |
XP_011528028.1 |
XM_011529727.2 |
182 |
Intron |
|
|
XP_011528029.1 |
XM_011529728.2 |
182 |
Intron |
|
|
XP_011528030.1 |
XM_011529729.2 |
182 |
Intron |
|
|
XP_011528031.1 |
XM_011529730.2 |
182 |
Missense Mutation |
CCG,CTG |
P7L |
XP_011528032.1 |
XM_017028454.1 |
182 |
Missense Mutation |
CCG,CTG |
P7L |
XP_016883943.1 |
XM_017028455.1 |
182 |
Intron |
|
|
XP_016883944.1 |
View Full Product Details