Product Details

SNP ID
rs202200507
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30541794 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGACCACCAAAGCCTCCACAGC[C/T]ATATCCCAGGCCTCTGTAGTAGCTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRTAP19-6 PubMed Links

Gene Details

Gene
KRTAP19-6
Gene Name
keratin associated protein 19-6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303120.1 71 Missense Mutation AGC,GGC S14G NP_001290049.1
NM_181612.3 71 Missense Mutation AGC,GGC S14G NP_853643.1

View Full Product Details