Product Details

SNP ID
rs202202057
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30501887 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAAGGATTTCAATAAAATCCAGA[G/T]AATCCATATCCTCCATAGCATGATG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KRTAP19-3 PubMed Links

Gene Details

Gene
KRTAP19-3
Gene Name
keratin associated protein 19-3
There are no transcripts associated with this gene.

Gene
KRTAP19-4
Gene Name
keratin associated protein 19-4
There are no transcripts associated with this gene.

Gene
KRTAP19-5
Gene Name
keratin associated protein 19-5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181611.1 204 Missense Mutation TTA,TTC L68F NP_853642.1

View Full Product Details