Product Details

SNP ID
rs199724044
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30881473 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTGGGCCACTGGAACAATGCAGC[A/G]TCCTCCAATGGGCCTGGAAGAGCAA
Phenotype
MIM: 600064
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRTAP11-1 PubMed Links

Gene Details

Gene
KRTAP11-1
Gene Name
keratin associated protein 11-1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175858.2 83 Missense Mutation CGC,TGC R18C NP_787054.1

View Full Product Details