Product Details

SNP ID
rs199733949
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:31038284 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGCAGAGAATCCATATCCTCCGT[A/G]GTATAATGGTCGGCAACAGCTGAAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRTAP19-8 PubMed Links

Gene Details

Gene
KRTAP19-8
Gene Name
keratin associated protein 19-8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099219.1 193 Missense Mutation CAC,TAC H54Y NP_001092689.1

View Full Product Details