Product Details

SNP ID
rs199813683
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:33505685 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCAAATTCTACACGATTTTTATA[C/T]AGTTTATGATTAATTACCTGTAATA
Phenotype
MIM: 138440
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GART PubMed Links

Gene Details

Gene
GART
Gene Name
phosphoribosylglycinamide formyltransferase, phosphoribosylglycinamide synthetase, phosphoribosylaminoimidazole synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000819.4 2912 Silent Mutation CTA,CTG L867L NP_000810.1
NM_001136005.1 2912 Silent Mutation CTA,CTG L867L NP_001129477.1
NM_001136006.1 2912 Silent Mutation CTA,CTG L867L NP_001129478.1
NM_175085.2 2912 Intron NP_780294.1
XM_005260941.1 2912 Silent Mutation CTA,CTG L867L XP_005260998.1
XM_006723989.1 2912 Silent Mutation CTA,CTG L867L XP_006724052.1
XM_006723990.1 2912 Silent Mutation CTA,CTG L867L XP_006724053.1
XM_011529526.1 2912 Silent Mutation CTA,CTG L867L XP_011527828.1

View Full Product Details