Product Details
- SNP ID
-
rs200613884
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:30813083 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTAGCCGAAAGCCCCACAGCCGTT[G/T]TAGCCATAGCCGAAGCCATAGCCCA
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KRTAP8-1
PubMed Links
Gene Details
- Gene
- KRTAP8-1
- Gene Name
- keratin associated protein 8-1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_175857.3 |
170 |
Nonsense Mutation |
TAA,TAC |
*46Y |
NP_787053.1 |
View Full Product Details