Product Details
- SNP ID
-
rs200618900
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:39406353 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTCTTCTTTGTCTGAGGGGGCCTTT[C/T]GTGTAGGGAGCAGTGCCTTTGCCAA
- Phenotype
-
MIM: 602915
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LCA5L
PubMed Links
Gene Details
- Gene
- LCA5L
- Gene Name
- LCA5L, lebercilin like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152505.3 |
1527 |
Silent Mutation |
ACA,ACG |
T514T |
NP_689718.1 |
XM_005260926.1 |
1527 |
Silent Mutation |
ACA,ACG |
T514T |
XP_005260983.1 |
XM_006723967.1 |
1527 |
Silent Mutation |
ACA,ACG |
T514T |
XP_006724030.1 |
XM_006723972.1 |
1527 |
Silent Mutation |
ACA,ACG |
T384T |
XP_006724035.1 |
XM_006723973.1 |
1527 |
Silent Mutation |
ACA,ACG |
T384T |
XP_006724036.1 |
XM_011529458.1 |
1527 |
Silent Mutation |
ACA,ACG |
T563T |
XP_011527760.1 |
XM_011529459.1 |
1527 |
Silent Mutation |
ACA,ACG |
T563T |
XP_011527761.1 |
XM_011529460.1 |
1527 |
Silent Mutation |
ACA,ACG |
T563T |
XP_011527762.1 |
XM_011529461.1 |
1527 |
Silent Mutation |
ACA,ACG |
T563T |
XP_011527763.1 |
XM_011529462.1 |
1527 |
Silent Mutation |
ACA,ACG |
T563T |
XP_011527764.1 |
XM_011529463.2 |
1527 |
Silent Mutation |
ACA,ACG |
T563T |
XP_011527765.1 |
XM_011529464.1 |
1527 |
Silent Mutation |
ACA,ACG |
T563T |
XP_011527766.1 |
XM_011529465.1 |
1527 |
Silent Mutation |
ACA,ACG |
T563T |
XP_011527767.1 |
XM_011529466.1 |
1527 |
Silent Mutation |
ACA,ACG |
T563T |
XP_011527768.1 |
XM_011529467.1 |
1527 |
Silent Mutation |
ACA,ACG |
T563T |
XP_011527769.1 |
XM_011529468.1 |
1527 |
Silent Mutation |
ACA,ACG |
T563T |
XP_011527770.1 |
XM_011529469.2 |
1527 |
Silent Mutation |
ACA,ACG |
T563T |
XP_011527771.1 |
XM_011529470.1 |
1527 |
Silent Mutation |
ACA,ACG |
T563T |
XP_011527772.1 |
XM_011529471.1 |
1527 |
Silent Mutation |
ACA,ACG |
T517T |
XP_011527773.1 |
XM_017028274.1 |
1527 |
Silent Mutation |
ACA,ACG |
T384T |
XP_016883763.1 |
XM_017028275.1 |
1527 |
Silent Mutation |
ACA,ACG |
T384T |
XP_016883764.1 |
XM_017028276.1 |
1527 |
Silent Mutation |
ACA,ACG |
T384T |
XP_016883765.1 |
XM_017028277.1 |
1527 |
Silent Mutation |
ACA,ACG |
T384T |
XP_016883766.1 |
XM_017028278.1 |
1527 |
Silent Mutation |
ACA,ACG |
T384T |
XP_016883767.1 |
- Gene
- WRB
- Gene Name
- tryptophan rich basic protein
There are no transcripts associated with this gene.
- Gene
- WRB-SH3BGR
- Gene Name
- WRB-SH3BGR readthrough
View Full Product Details